Learn More
IKA™ MX.PP Sample Mixers
MX.PP - Set of 3 sample mixers made of polypropylene, each with a different mixing type.
Brand:  IKA™ 0020103092
Product Code. 17573636
  • £71.70 / Each
Estimated Shipment: 11-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Set of 3 sample mixers made of polypropylene, each with a different mixing type.
  • The package includes:
    • T shape mixing chip * 1
    • Y shape mixing chip * 1
    • Interrupted flow mixing chip * 1.
    • All three mixing chips are made of polypropylene (PP).
  • Scope of delivery
    • T shape mixing chip, PP
    • Y shape mixing chip, PP
    • Interrupted flow mixing chip, PP
Specifications
Specifications
Polypropylene (PP)
T Shape Mixing Chip, Y Shape Mixing Chip, Interrupted Flow Mixing Chip