missing translation for 'onlineSavingsMsg'
Learn More

MYC Rabbit anti-Human, Clone: 2B17, Proteintech

Rabbit Recombinant Antibody

Brand:  Proteintech 80845-1-RR-100UL

 View more versions of this product

Product Code. 17679796

  • £360.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

MYC antibody apecifically detetects MYC antigen in Human Samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

MYC
2B17
Unconjugated
BC000141
Rabbit
Protein A purification
Primary
Human
Liquid
Western Blot, ELISA
1000 μg/mL
PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
MYC
MYC fusion protein Ag1263
100 μL
4609
Store at -20°C. Stable for one year after shipment.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers