missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lonza™ MycoAlert™ PLUS Mycoplasma Detection Kit (100 Tests)
Detect mycoplasma contamination in cell culture supernatants, media, and supplements with the next generation MycoAlertTM Kit allowing the use of less sensitive plate luminometers or multifunctional readers.
Brand: Lonza™ LT07-710
This item is not returnable.
View return policy
Description
- Easy assay: Simply add two reagents to the culture supernatant and perform two luminescence readings.
- Results in < 20 min.
- Easy interpretation of results.
- Detects all common mollicute contaminations (Mycoplasma, Acholeplasma, Entomoplasma and Spiroplasma; except Ureaplasma).
- Suited for tube luminometers, plate luminometers and multifunctional readers.
- Next-generation MycoAlert™ Kit which provides higher light output, facilitating use with less sensitive plate luminometers or multifunctional readers as well as testing of unused media, media supplements, or water.
Specifications
Light yellow | |
Mycoplasma detection | |
1 x 10 mL MycoALert™ PLUS Reagent (LyophiLized), 1 x 10 mL MycoALert™ PLUS Substrate (LyophiLized), 1 x 20 mL MycoALert™ PLUS Assay Buffer |
MycoAlert™ PLUS Reagent (lyophilized), MycoAlert™ PLUS Substrate, MycoAlert™ PLUS Assay Buffer | |
7.5 |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- MycoAlert PLUS Mycoplasma Detection Kit
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
Precautionary Statement
- P264-Wash skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : N-glycylglycine
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction