missing translation for 'onlineSavingsMsg'
Learn More

PEPperCHIP™ Mycobacterium Tuberculosis Antigen Microarray

Mycobacterium Tuberculosis Antigen Microarray with 3751 antigen specific peptides printed in duplicate. The microarray can be used for epitope mapping, serum screening or biomarker discovery.

Brand:  PEPperCHIP™ SPC.018.001

Additional Details : Weight : 0.02200kg

Product Code. 16307768

  • £482.30 / Each
Estimated Shipment: 22-10-2024
to see stock.
Add to basket

Description

Description

  • Microarray Content: 13 selected antigens of mycobacterium tuberculosis: ESX-1 secretion-associated protein, espB , antigen 85-A, fbpA, ESAT-6-like protein, esxB, alanine and proline-rich secreted protein, apa, lipoprotein, lprG, alpha-crystallin, hspX, phosphate-binding protein, pstS1, ESX-1 secretion-associated protein, espA, ESX-1 secretion-associated protein, espE, immunogenic protein, mpt64, enoyl-CoA hydratase, echA3 , probable cutinase, cfp21, cell surface lipoprotein, mpt83
  • Peptide Length/Overlap: 15 aa/ 14 aa
  • Number of Peptides: 3751 printed in duplicate and HA, polio and c-Myc control peptides
Specifications

Specifications

PEPperSlide™ PEGMA/PMMA
Antibody validation, epitope mapping, serum screening, biomarker discovery
Microscopic Glass
Individual barcode for each microarray
1 Slide
1 mm
Transparent
75.4 x 25 mm
High-density Peptide Microarray
Non-sterile
1 array copy per microarray
Combinatorial peptide synthesis by amino acid laser printing
ISO 9001:2015
RUO
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers