missing translation for 'onlineSavingsMsg'
Learn More

Mycophenolic acid-d3, MedChemExpress

Product Code. 16400928
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
This item is not returnable. View return policy

Product Code. 16400928

Brand: Medchem Express HYB0421S

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Mycophenolic acid D3 (Mycophenolate D3) is deuterium labeled Mycophenolic acid, which is an an immunosuppresant drug and has potent anti-proliferative activity.

Mycophenolic acid D3 (Mycophenolate D3) is deuterium labeled Mycophenolic acid, which is an an immunosuppresant drug and has potent anti-proliferative activity.

Specifications

Chemical Name or Material Mycophenolic acid-d3
CAS 1185242-90-3
Purity Grade Notes Research
Physical Form Solid
Molecular Formula C17H17D3O6
Color White
Synonym Mycophenolate-d3
SMILES O=C(OC1)C2=C1C(C)=C(OC([2H])([2H])[2H])C(C/C=C(C)/CCC(O)=O)=C2O
Molecular Weight (g/mol) 323.36
Quantity 5 mg
Formula Weight 323.36
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.