Learn More
Lonza™ MycoZap™ Plus-PR
Antibiotic formulation for prevention of bacteria, fungi, and mycoplasma contaminations in primary cell cultures.
Brand:  Lonza™ VZA-2021
Product Code. 11566529
  • £313.00 / Pack of 10
Explore more special offers
Description
Description
  • Active against mycoplasma, Gram negative and Gram positive bacteria, as well as yeast and fungi.
  • Complete solution replacing Pen/Strep formulation.
  • Applications:
    • Optimized for gentle protection of primary cells.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Prevention of cell culture contamination
4.5 at 20°C
10 x 1 mL