missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myeloma Proteins IgG2a(Kappa), [UPC 10] Isotype Control, MP Biomedicals
Mouse Monoclonal Antibody
Brand: MP Biomedicals 0850328
Additional Details : Weight : 0.00010kg
Description
Myeloma Proteins IgG2a(Kappa) Monoclonal antibody specifically detects Myeloma Proteins IgG2a(Kappa) in samples. It is validated for ELISA.Specifications
IgG2a κ | |
Monoclonal | |
Unconjugated | |
Mouse | |
1 mg |
ELISA | |
UPC 10 | |
Purified proteins are dialyzed into 0.01M Sodium Phosphate, 0.13M sodium chloride, pH 7.2 | |
0.22μm filtration | |
Store unopened product at -70°C. Store opened material no longer than a few days at 2° to 8°C unless sodium azide is added. For long-term storage, aliquot and freeze at -70°C. |