missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYH9, Mouse anti-Human, Clone: 4H3, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00004627-M06.100ug
This item is not returnable.
View return policy
Description
Sequence: RLKQLKRQLEEAEEEAQRANASRRKLQRELEDATETADAMNREVSSLKNKLRRGDLPFVVPRRMARKGAGDGSDEEVDGKADGAEAKPAESpecifications
MYH9 | |
Monoclonal | |
Unconjugated | |
myosin, heavy chain 9, non-muscle | |
DFNA17/EPSTS/FTNS/MGC104539/MHA/NMHC-II-A/NMMHCA | |
Mouse | |
100 μg | |
Primary | |
Human | |
Antibody |
ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
4H3 | |
In 1x PBS, pH 7.4 | |
NM_002473.5 | |
MYH9 | |
MYH9 (NP_002464.1, 1871 a.a. ∼ 1960 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
RUO | |
4627 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG2b κ |