missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals myo-Inositol, Cell Culture Grade Product Code.: 15495179

Thermo Scientific Chemicals myo-Inositol, Cell Culture Grade

Product Code. 15495179
50 g
Click to view available options
Quantity:
50 g
100 g
250 g

Product Code. 15495179

Brand: Thermo Scientific Chemicals J62886.18

Please to purchase this item. Need a web account? Register with us today!

A lipotropic agent

It has been tested with CHO, L929, and Vero cell lines and can be used for other cell culture applications. A method for HPLC identification and quantitation has been achieved by this chemical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It has been tested with CHO, L929, and Vero cell lines and can be used for other cell culture applications. A method for HPLC identification and quantitation has been achieved by this chemical.

Solubility
It is soluble in water (50 mg/ml).

Notes
Solutions of myo-inositol can be autoclaved for use in cell culture. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87-89-8
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.156
MDL Number MFCD00077932
InChI Key CDAISMWEOUEBRE-UHFFFAOYSA-N
Synonym scyllo-inositol, myo-inositol, inositol, muco-inositol, epi-inositol, i-inositol, meso-inositol, allo-inositol, 1d-chiro-inositol, myoinositol
PubChem CID 892
ChEBI CHEBI:24848
IUPAC Name cyclohexane-1,2,3,4,5,6-hexol
SMILES C1(C(C(C(C(C1O)O)O)O)O)O

Specifications

Melting Point 222°C to 227°C
Color White
Odor Odorless
Quantity 50 g
Beilstein 1907329
Merck Index 14,4978
Solubility Information It is soluble in water (50mg/ml).
Formula Weight 180.16
Grade Cell Culture
Physical Form Crystalline Powder
Chemical Name or Material myo-Inositol
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberY02J027 Certificate TypeCertificate of Analysis Date13/11/2023 Catalog NumberJ62886.22,J62886.30,J62886.18
Lot NumberX27J025 Certificate TypeCertificate of Analysis Date03/11/2023 Catalog NumberJ62886.22,J62886.30,J62886.18
Lot NumberX27J026 Certificate TypeCertificate of Analysis Date03/11/2023 Catalog NumberJ62886.22,J62886.30,J62886.18
Lot NumberS08J046 Certificate TypeCertificate of Analysis Date25/10/2023 Catalog NumberJ62886.22,J62886.30,J62886.18
Lot NumberU15J015 Certificate TypeCertificate of Analysis Date14/09/2023 Catalog NumberJ62886.22,J62886.30,J62886.18

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals myo-Inositol, Cell Culture Grade >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.