missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYOM3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17692-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody recognizes the intact MYOM3 protein (160-160 kDa) as well as MYOM3 fragments (100 kDa and 130 kDa).
MYOM3 (myomesin 3) is a structural component of the M-band in striated muscle and is involved in sarcomere stability and resistance during intense or sustained stretching. MYOM3 can be detected mainly in intermediate speed fibers of skeletal muscle. Recently high level of MYOM3 fragments were detected in sera from patients with muscular dystrophy, including Duchenne muscular dystrophy (DMD). MYOM3 fragments may be used as serum biomarker for DMD and other neuromuscular disorders.Specifications
MYOM3 | |
Polyclonal | |
Unconjugated | |
MYOM3 | |
MYOM3, Myomesin 3, Myomesin family member 3, myomesin family, member 3, RP11 293P20.1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
127294, 242702 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
A2ABU4, Q5VTT5 | |
Myom3 | |
MYOM3 Fusion Protein Ag11849 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |