Learn More
Myosin 4 Monoclonal Antibody (MF20), eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£120.00 - £292.00
Specifications
Antigen | Myosin 4 |
---|---|
Clone | MF20 |
Concentration | 0.5 mg/mL |
Applications | Immunocytochemistry, Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15597456
|
Invitrogen
14-6503-80 |
25 μg |
£120.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15507466
|
Invitrogen
14-6503-82 |
100 μg |
£292.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Description: This MF20 monoclonal antibody recognizes the heavy chain of myosin II, specificially the light meromyosin portion, in cardiac and skeletal muscle of vertebrates. Myosin II is composed of two heavy chains and four light chains. The 220-kDa myosin heavy chain exists as four different isoforms due to alternative splicing. Myosins interact with actin and hydrolyze ATP to function in muscle contraction, cytokinesis, and phagocytosis. The MF20 has been shown to react to myosin from a variety of mammalian, avian and amphibian species, including rat, mouse, human, chicken, zebrafish, and dog. Applications Reported: This MF20 antibody has been reported for use in western blotting, and immunocytochemistry. Applications Tested: This MF20 antibody has been tested by immunocytochemistry and western blot and can be used at less than or equal to 1 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.
Target protein includes a fast myosin isoform expressed in skeletal muscle and has function involved in Muscle contraction.Specifications
Myosin 4 | |
0.5 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
2B myosin heavy chain; AI506973; fast myosin heavy chain 2B; LOW QUALITY PROTEIN: myosin-4; MHC; MHC2B; Minimsc; Minmus; MM; MYH1; MYH2B; MYH-2B; myh4; MyHC IIb isoform; myHC-2b; MyHC-IIb; Myhsf; myosin heavy chain; myosin heavy chain 1; myosin heavy chain 2b; myosin heavy chain 2X; myosin heavy chain 4; Myosin heavy chain IIb; myosin heavy chain IIb isoform; myosin heavy chain isoform 2b; myosin heavy chain type IIb; Myosin heavy chain, skeletal muscle; myosin heavy chain, skeletal muscle, fetal; Myosin heavy chain, skeletal muscle, juvenile; myosin, heavy chain 4, skeletal muscle; myosin, heavy polypeptide 4, skeletal muscle; myosin-4; unnamed protein product | |
MYH4 | |
Primary | |
4° C | |
MYH4 |
MF20 | |
Immunocytochemistry, Western Blot | |
Unconjugated | |
Mouse | |
Bovine, Canine, Chicken, Chimpanzee, Feline, Guinea Pig, Human, Mouse, Non-human Primate, Rabbit, Rat | |
Q076A5, Q28641, Q29RW1, Q5SX39, Q9Y623 | |
100009548, 100125991, 100726456, 17884, 360543, 408020, 4622, 468396, 479502 | |
IgG2b κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.