missing translation for 'onlineSavingsMsg'
Learn More

Gilson™ MyPIPETMAN™ Select 4 Pipette Kit, P2, P20, P200, P1000

The MyPIPETMAN™ Select 4 Pipette Kit includes pipettes with reinforced ergonomics for optimal use in many protocols and most liquid handling tasks in molecular biology, immunology and cell culture. Included are the most commonly used pipettes: P2, P20, P200, and P1000; plus tips and accessories.

Brand:  Gilson™ F167390

1262.84 GBP valid until 2025-04-30
Use promo code "24138" to get your promotional price.


Product Code. 17838027

  • £1365.00 / 1 set
Estimated Shipment: 24-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

MyPIPETMAN™ Select 4-Pipette Kit includes extra customization accessories (colored windows) are also provided to identify the use of the pipettes for a specific liquid, room, or technique.

  • Simply adjust the volume by turning the push button of the pipette with the unique, patented Trilock™ volume-locking system
  • Free volume setting position for quick and effortless volume adjustment
  • Light ratchet volume setting for precise volume adjustment
  • Volume locked position to secure pipetting
  • Lightweight and well-balanced, MyPIPETMAN™ Select has an elegant design and an optimized grip
  • Built-in ejection system provides reduced tip ejection forces for convenient use throughout the day
  • Fully autoclavable without disassembly for maximum convenience and protection from cross-contamination
  • Autoclavable plastic tip ejectors, as extra accessories, allow using the pipettes when handling corrosive liquids
  • Pipettes have a longer life span and can be used with different kinds of liquids without any risk of damaging the metal tip ejectors
  • SINGLE™ pipette holders are also provided in each kit to store each pipette vertically on a wall, protecting the pipette from contamination without taking up any precious bench space
  • Includes: 4 MyPIPETMAN™ Select pipettes: P2, P20, P200, and P1000 equipped with metal ejectors, 3 PIPETMAN™ DIAMOND Tips TIPACKS (models: DL10, D200 and D1000), 4 SINGLE pipette holders, 4 white plastic tip ejectors, 6 colored windows (red, yellow, blue, green, white, and dark blue)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Yes
Blue/White
Universal Pipette Tip
Polypropylene, Stainless Steel
1
Non-sterile
1000 μL
PCR Ready
Autoclavable
Reusable
Ergonomic
P2, P20, P200, P1000
Pipetman
Pipette Kit
1 kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gilson™ MyPIPETMAN™ Select 4 Pipette Kit, P2, P20, P200, P1000 > Starter Kit; Color: Blue/White

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.