missing translation for 'onlineSavingsMsg'
Learn More

Myricetin, 98%, Thermo Scientific Chemicals

Product Code. 15423219
Click to view available options
Quantity:
10 mg
25 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Myricetin is used as an antioxidant. It is a cell-permeable flavonoid used in inflammatory, diabetes, and cancer studies. It is effective in protecting neurons against oxidative stresses. Further, it inhibits yeast alfa-glucosidase,1 glyoxalase I in vitro and bovine milk xanthine oxidase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Myricetin is used as an antioxidant. It is a cell-permeable flavonoid used in inflammatory, diabetes, and cancer studies. It is effective in protecting neurons against oxidative stresses. Further, it inhibits yeast alfa-glucosidase,1 glyoxalase I in vitro and bovine milk xanthine oxidase.

Solubility
Soluble in dimethyl sulfoxide, dimethyl formamide and ethanol. Insoluble in water.  

Notes
Store in a cool place. Incompatible with strong oxidizing agents.    
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 529-44-2
Molecular Formula C15H10O8
Molecular Weight (g/mol) 318.24
MDL Number MFCD00006827
InChI Key IKMDFBPHZNJCSN-UHFFFAOYSA-N
Synonym myricetin, cannabiscetin, myricetol, myricitin, 3,5,7-trihydroxy-2-3,4,5-trihydroxyphenyl-4h-chromen-4-one, 3,3',4',5,5',7-hexahydroxyflavone, 3,5,7,3',4',5'-hexahydroxyflavone, unii-76xc01ftoj, ccris 5838, 3,5,7-trihydroxy-2-3,4,5-trihydroxyphenyl-4h-1-benzopyran-4-one
PubChem CID 5281672
ChEBI CHEBI:18152
SMILES OC1=CC(O)=C2C(OC(=C(O)C2=O)C2=CC(O)=C(O)C(O)=C2)=C1

Specifications

Melting Point >300°C
Color Brown-Yellow
Quantity 10 mg
Beilstein 332331
Merck Index 14,6332
Solubility Information Soluble in dimethyl sulfoxide, dimethyl formamide and ethanol; Insoluble in water .
IUPAC Name 3,5,7-trihydroxy-2-(3,4,5-trihydroxyphenyl)chromen-4-one
Formula Weight 318.24
Percent Purity 98%
Physical Form Crystalline Solid
Chemical Name or Material Myricetin
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'