Learn More
Myristoyl chloride, 97%, Thermo Scientific Chemicals
£18.30 - £236.00
Chemical Identifiers
CAS | 112-64-1 |
---|---|
Molecular Formula | C14H27ClO |
Molecular Weight (g/mol) | 246.82 |
MDL Number | MFCD00000741 |
InChI Key | LPWCRLGKYWVLHQ-UHFFFAOYSA-N |
Synonym | myristoyl chloride, myristoylchloride, tetradecanoylchloride, e r eao poundae, myristic acid chloride, tetradecanoic acid chloride, acmc-2099fr, ksc181q3r |
PubChem CID | 66986 |
IUPAC Name | tetradecanoyl chloride |
SMILES | CCCCCCCCCCCCCC(=O)Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10635062
|
Thermo Scientific Acros
180630050 |
5 g | Glass bottle |
£18.30
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10458930
|
Thermo Scientific Acros
180631000 |
100 g | Glass bottle |
£60.40
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10157803
|
Thermo Scientific Acros
180635000 |
500 g | Glass bottle |
£236.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
112-64-1 | |
246.82 | |
LPWCRLGKYWVLHQ-UHFFFAOYSA-N | |
66986 | |
CCCCCCCCCCCCCC(=O)Cl |
C14H27ClO | |
MFCD00000741 | |
myristoyl chloride, myristoylchloride, tetradecanoylchloride, e r eao poundae, myristic acid chloride, tetradecanoic acid chloride, acmc-2099fr, ksc181q3r | |
tetradecanoyl chloride |
Specifications
112-64-1 | |
100.0 | |
172°C to 176°C (5.0 mbar) | |
Authentic | |
C14H27ClO | |
1.4480 to 1.45 | |
02, 368 | |
Solubility in water: may decompose | |
CCCCCCCCCCCCCC(=O)Cl | |
246.82 | |
246.82 | |
Liquid |
96.0 | |
-1°C | |
>100°C | |
96% min. (GC) | |
CH3(CH2)12COCl | |
MFCD00000741 | |
myristoyl chloride, myristoylchloride, tetradecanoylchloride, e r eao poundae, myristic acid chloride, tetradecanoic acid chloride, acmc-2099fr, ksc181q3r | |
LPWCRLGKYWVLHQ-UHFFFAOYSA-N | |
tetradecanoyl chloride | |
66986 | |
97% | |
Myristoyl chloride |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful if inhaled.
May form explosive peroxides.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 203-994-6
- Myristoyl chloride
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
- EUH029-Contact with water liberates toxic gas.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only