Learn More
Myristyltrimethylammonium bromide, 99%, Thermo Scientific Chemicals
£20.40 - £593.96
Chemical Identifiers
CAS | 1119-97-7 |
---|---|
Molecular Formula | C17H38BrN |
Molecular Weight (g/mol) | 336.40 |
MDL Number | MFCD00011770 |
InChI Key | CXRFDZFCGOPDTD-UHFFFAOYSA-M |
Synonym | cetrimide, tetradecyltrimethylammonium bromide, myristyltrimethylammonium bromide, tetradonium bromide, n,n,n-trimethyltetradecan-1-aminium bromide, tetradecyl trimethyl ammonium bromide, myrtrimonium bromide, mytab, mitmab, morpan t |
PubChem CID | 14250 |
ChEBI | CHEBI:3565 |
IUPAC Name | trimethyl(tetradecyl)azanium;bromide |
SMILES | [Br-].CCCCCCCCCCCCCC[N+](C)(C)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10439930
|
Thermo Scientific Acros
229370050 |
5 g | Glass bottle |
£20.40
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10266570
|
Thermo Scientific Acros
229371000 |
100 g | Plastic bottle |
£52.36
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10582821
|
Thermo Scientific Acros
229375000 |
500 g | Plastic bottle |
£140.70
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10501794
|
Thermo Scientific Acros
229370025 |
2.5 kg | Plastic bottle |
£593.96
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalystChemical Identifiers
1119-97-7 | |
336.40 | |
CXRFDZFCGOPDTD-UHFFFAOYSA-M | |
14250 | |
trimethyl(tetradecyl)azanium;bromide |
C17H38BrN | |
MFCD00011770 | |
cetrimide, tetradecyltrimethylammonium bromide, myristyltrimethylammonium bromide, tetradonium bromide, n,n,n-trimethyltetradecan-1-aminium bromide, tetradecyl trimethyl ammonium bromide, myrtrimonium bromide, mytab, mitmab, morpan t | |
CHEBI:3565 | |
[Br-].CCCCCCCCCCCCCC[N+](C)(C)C |
Specifications
1119-97-7 | |
100.0 | |
White | |
Authentic | |
C17H38BrN | |
MFCD00011770 | |
cetrimide, tetradecyltrimethylammonium bromide, myristyltrimethylammonium bromide, tetradonium bromide, n,n,n-trimethyltetradecan-1-aminium bromide, tetradecyl trimethyl ammonium bromide, myrtrimonium bromide, mytab, mitmab, morpan t | |
CXRFDZFCGOPDTD-UHFFFAOYSA-M | |
trimethyl(tetradecyl)azanium;bromide | |
14250 | |
336.39 | |
Fine Crystalline Powder |
98.5 | |
245°C to 250°C | |
6 to 8 (1% aq. soln.) | |
98.5 to 101.5% (Argentometry) | |
CH3(CH2)13N(CH3)3Br | |
15,6422 | |
Solubility in water: 100g/L (20°C) | |
[Br-].CCCCCCCCCCCCCC[N+](C)(C)C | |
336.40 | |
CHEBI:3565 | |
99% | |
Myristyltrimethylammonium bromide, 99% |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 214-291-9
missing translation for 'rtecsNumber' : BS5776000
missing translation for 'tsca' : TSCA
- Myristyltrimethylammonium bromide
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H335-May cause respiratory irritation.
- H373-May cause damage to organs.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
RUO – Research Use Only