Learn More
Thermo Scientific™ mySPIN™ 6 Mini Centrifuge
Our most compact centrifuge, the mySPIN 6 mini centrifuge, is ideal for microfilter cell separations and HPLC samples, with adapters to accommodate all major microcentrifuge tubes.
Brand: Thermo Scientific™ 75004087
43.24 GBP valid until 2024-12-31
Use promo code "21929" to get your promotional price.
Description
Bring efficiency and user comfort to your lab stations with the Thermo Scientific mySPIN 6 mini centrifuge. Ergonomic design, tool-free rotors, and quick-spin options make it fast and easy to use. Ideal for microfilter cell separations and HPLC samples, with adapters to accommodate all major microtube and PCR tube styles (2.0, 1.5, 0.5, 0.2 mL) for customized use.
Enjoy convenience at your personal workstation with our most compact centrifuges, featuring:- Small footprint for efficient use of lab space
- Easy operation and palm-shaped lid for left- or right-handed users
- Quick-spin feature
- Quiet operation, low vibration
- Safety features including unit will not operate without lid in place for safety
- Includes 2 rotors to accommodate 0.2, 0.5, 1.5, 2.0 mL microcentrifuge tubes and PCR strip tubes
- Tool-free, quick rotor exchange
Specifications
mySPIN6 Adapter | |
12 V |