missing translation for 'onlineSavingsMsg'
Learn More
n-(1,3-benzodioxol-5-ylmethyl)-n-methylamine, 97%, Thermo Scientific™ Product Code.: 10796144

n-(1,3-benzodioxol-5-ylmethyl)-n-methylamine, 97%, Thermo Scientific™

Product Code. 10796144
1 g
Click to view available options
Quantity:
250 mg
1 g
10 g
This item is not returnable. View return policy

Product Code. 10796144

Brand: Thermo Scientific Maybridge CC01646DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 15205-27-3
Molecular Formula C9H12NO2
Molecular Weight (g/mol) 166.20
MDL Number MFCD04496422
InChI Key CEPGPPSMCRKGFJ-UHFFFAOYSA-O
Synonym n-1,3-benzodioxol-5-ylmethyl-n-methylamine, 1-1,3-benzodioxol-5-yl-n-methylmethanamine, 2h-1,3-benzodioxol-5-ylmethyl methyl amine, n-methyl-3,4-methylenedioxy benzylamine, 1,3-benzodioxole-5-methanamine,n-methyl, 1,3-benzodioxole-5-methanamine, n-methyl, 2h-1,3-benzodioxol-5-yl methyl methyl amine, 1,3-benzodioxol-5-ylmethyl methylaminehydrochloride, n-methylpiperonylamine, 3,4-methylenedioxy-n-methylbenzylamineShow MoreShow Less
PubChem CID 421238
SMILES C[NH2+]CC1=CC=C2OCOC2=C1

Specifications

Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
IUPAC Name 1-(1,3-benzodioxol-5-yl)-N-methylmethanamine
Formula Weight 165.19
Percent Purity 97%
Chemical Name or Material N-(1, 3-Benzodioxol-5-ylmethyl)-N-methylamine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
n-(1,3-benzodioxol-5-ylmethyl)-n-methylamine, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.