missing translation for 'onlineSavingsMsg'
Learn More
N-(1-Benzothien-2-ylmethyl)-N-methylamine hydrochloride, ≥97%, Thermo Scientific™ Product Code.: 10780334

N-(1-Benzothien-2-ylmethyl)-N-methylamine hydrochloride, ≥97%, Thermo Scientific™

Product Code. 10780334
250 mg
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy

Product Code. 10780334

Brand: Thermo Scientific Maybridge CC12296CB

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 849776-43-8
Molecular Formula C10H12ClNS
Molecular Weight (g/mol) 213.72
MDL Number MFCD03306023
InChI Key PBEZSHRKTUDPLK-UHFFFAOYSA-N
Synonym n-1-benzothien-2-ylmethyl-n-methylamine hydrochloride, 1-benzo b thiophen-2-yl-n-methylmethanamine hydrochloride, 1-benzothien-2-ylmethyl methylamine hydrochloride, 1-benzothiophen-2-ylmethyl methyl amine hydrochloride, benzo b thiophen-2-ylmethyl-methyl-ammonium chloride, c10h11ns.hcl, benzo b thiophen-2-ylmethyl methylamine, chloride, 1-benzo b thiophen-2-yl ;-n-methylmethanamine hydrochloride, 1-1-benzothiophen-2-yl-n-methylmethanamine-hydrogen chloride 1/1
PubChem CID 16956076
SMILES Cl.CNCC1=CC2=CC=CC=C2S1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
IUPAC Name [(1-benzothiophen-2-yl)methyl](methyl)amine hydrochloride
Formula Weight 213.73
Percent Purity 97%
Chemical Name or Material N-(1-Benzothien-2-ylmethyl)-N-methylamine hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N-(1-Benzothien-2-ylmethyl)-N-methylamine hydrochloride, ≥97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.