missing translation for 'onlineSavingsMsg'
Learn More

N-(1-Naphthyl)-3-nitrobenzenesulfonamide, 97%, Thermo Scientific™

Product Code. 15444838
Click to view available options
Quantity:
100 mg
500 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 419559-79-8
Molecular Formula C16H12N2O4S
Molecular Weight (g/mol) 328.34
MDL Number MFCD01136840
InChI Key WIFWFLHGNPQTSE-UHFFFAOYSA-N
Synonym n-1-naphthyl-3-nitrobenzenesulfonamide, n-naphthalen-1-yl-3-nitrobenzenesulfonamide, n-1-naphthyl-m-nitrobenzenesulfonamide, n-1-naphthyl-3-nitrobenzenesulfonamide #
PubChem CID 232046
IUPAC Name N-naphthalen-1-yl-3-nitrobenzenesulfonamide
SMILES [O-][N+](=O)C1=CC(=CC=C1)S(=O)(=O)NC1=CC=CC2=CC=CC=C12

Specifications

Color Gray
Quantity 500 mg
Beilstein 2768935
Formula Weight 328.34
Percent Purity 97%
Physical Form Solid
Chemical Name or Material N-(1-Naphthyl)-3-nitrobenzenesulfonamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'