missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(1-Naphthyl)ethylenediamine dihydrochloride, ACS reagent, ≥98%, Honeywell Fluka™
Click to view available options
Quantity:
5 g
25 g
Specifications
CAS Max % | 1 |
Melting Point | 194°C to 198°C (Decomposition) |
Color | Colorless |
Quantity | 25 g |
Grade | ACS Grade Organics |
Chemical Name or Material | N-(1-Naphthyl)ethylenediamine dihydrochloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'