Learn More
N-(1-Naphthyl)ethylenediamine dihydrochloride, ACS, Thermo Scientific Chemicals
Used for colorimetric determination of sulfonamide in blood
£26.90 - £227.00
Chemical Identifiers
CAS | 1465-25-4 |
---|---|
Molecular Formula | C12H16Cl2N2 |
Molecular Weight (g/mol) | 259.174 |
MDL Number | MFCD00012556 |
InChI Key | MZNYWPRCVDMOJG-UHFFFAOYSA-N |
Synonym | n-1-naphthyl ethylenediamine dihydrochloride, n1-naphthalen-1-yl ethane-1,2-diamine dihydrochloride, marshall's reagent, n-1-naphthylethylenediamine dihydrochloride, ccris 425, unii-h734599kjl, 1,2-ethanediamine, n-1-naphthalenyl-, dihydrochloride, 2-1-naphthylamino ethylamine 2hcl, bratton-marshall reagent, n-1-naphthalenyl-1,2-ethanediamine dihydrochloride |
PubChem CID | 15106 |
ChEBI | CHEBI:53452 |
IUPAC Name | N'-naphthalen-1-ylethane-1,2-diamine;dihydrochloride |
SMILES | C1=CC=C2C(=C1)C=CC=C2NCCN.Cl.Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15455469
|
Thermo Scientific Alfa Aesar
J63214.06 |
5 g |
£26.90
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15470297
|
Thermo Scientific Alfa Aesar
J63214.14 |
25 g |
£91.10
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15480297
|
Thermo Scientific Alfa Aesar
J63214.22 |
100 g |
£227.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Used for colorimetric determination of sulfonamide in bloodN-(1-Naphthyl)ethylenediamine dihydrochloride acts as a coupling agent for the spectrophotometric determination of aromatic amines, aminophenols, phenylenediamines, dinitroanilines, chloroanilines, thiols and sulfonamides. It is also used in the quantitative analysis of nitrate and nitrite in water samples by colorimetry. It is involved in the preparation of (N-1-naphthyl-ethylenediamine)-dichloroplatinum(II) by reacting with potassium tetrachloroplatinate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsUsed for colorimetric determination of sulfonamide in bloodN-(1-Naphthyl)ethylenediamine dihydrochloride acts as a coupling agent for the spectrophotometric determination of aromatic amines, aminophenols, phenylenediamines, dinitroanilines, chloroanilines, thiols and sulfonamides. It is also used in the quantitative analysis of nitrate and nitrite in water samples by colorimetry. It is involved in the preparation of (N-1-naphthyl-ethylenediamine)-dichloroplatinum(II) by reacting with potassium tetrachloroplatinate.
Solubility
Soluble in water or 95% ethanol.Soluble in hot water, alcohol, dilute hydrochloric acid and acetone.
Notes
Hygroscopic. Light sensitive. Incompatible with acids, acid chlorides, acid anhydrides, halogen acids and oxidizing agents.
Chemical Identifiers
1465-25-4 | |
259.174 | |
MZNYWPRCVDMOJG-UHFFFAOYSA-N | |
15106 | |
N'-naphthalen-1-ylethane-1,2-diamine;dihydrochloride |
C12H16Cl2N2 | |
MFCD00012556 | |
n-1-naphthyl ethylenediamine dihydrochloride, n1-naphthalen-1-yl ethane-1,2-diamine dihydrochloride, marshall's reagent, n-1-naphthylethylenediamine dihydrochloride, ccris 425, unii-h734599kjl, 1,2-ethanediamine, n-1-naphthalenyl-, dihydrochloride, 2-1-naphthylamino ethylamine 2hcl, bratton-marshall reagent, n-1-naphthalenyl-1,2-ethanediamine dihydrochloride | |
CHEBI:53452 | |
C1=CC=C2C(=C1)C=CC=C2NCCN.Cl.Cl |
Specifications
1465-25-4 | |
Cream to White or Gray | |
MFCD00012556 | |
Hygroscopic; Light sensitive | |
n-1-naphthyl ethylenediamine dihydrochloride, n1-naphthalen-1-yl ethane-1,2-diamine dihydrochloride, marshall's reagent, n-1-naphthylethylenediamine dihydrochloride, ccris 425, unii-h734599kjl, 1,2-ethanediamine, n-1-naphthalenyl-, dihydrochloride, 2-1-naphthylamino ethylamine 2hcl, bratton-marshall reagent, n-1-naphthalenyl-1,2-ethanediamine dihydrochloride | |
MZNYWPRCVDMOJG-UHFFFAOYSA-N | |
N'-naphthalen-1-ylethane-1,2-diamine;dihydrochloride | |
15106 | |
259.18 | |
Powder |
191°C to 199°C | |
C12H16Cl2N2 | |
3707471 | |
14,6409 | |
Soluble in water or 95% ethanol;Soluble in hot water,alcohol,dilute hydrochloric acid and acetone. | |
C1=CC=C2C(=C1)C=CC=C2NCCN.Cl.Cl | |
259.174 | |
CHEBI:53452 | |
ACS | |
N-(1-Naphthyl)ethylenediamine dihydrochloride |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 215-981-2
missing translation for 'rtecsNumber' : KV5330000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only