missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(2,4,6-Trimethylphenyl)thiourea, 99%, Thermo Scientific™
£73.42 - £234.00
Chemical Identifiers
CAS | 91059-97-1 |
---|---|
Molecular Formula | C10H14N2S |
Molecular Weight (g/mol) | 194.296 |
MDL Number | MFCD00051488 |
InChI Key | LBUGQLLKHAHCJJ-UHFFFAOYSA-N |
Synonym | n-2,4,6-trimethylphenyl thiourea, n-mesitylthiourea, 2,4,6-trimethylphenyl thiourea, 1-2,4,6-trimethylphenyl thiourea, 1-2,4,6-trimethylphenyl-2-thiourea, 2,4,6-trimethyl-phenyl-thiourea, amino 2,4,6-trimethylphenyl amino methane-1-thione, 1-mesitylthiourea, acmc-20akda, maybridge1_006982 |
PubChem CID | 2731118 |
IUPAC Name | (2,4,6-trimethylphenyl)thiourea |
SMILES | CC1=CC(=C(C(=C1)C)NC(=S)N)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11489992
|
Thermo Scientific Alfa Aesar
A12083.14 |
25 g |
£73.42
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11499992
|
Thermo Scientific Alfa Aesar
A12083.22 |
100 g | N/A | ||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
91059-97-1 | |
194.296 | |
LBUGQLLKHAHCJJ-UHFFFAOYSA-N | |
2731118 | |
CC1=CC(=C(C(=C1)C)NC(=S)N)C |
C10H14N2S | |
MFCD00051488 | |
n-2,4,6-trimethylphenyl thiourea, n-mesitylthiourea, 2,4,6-trimethylphenyl thiourea, 1-2,4,6-trimethylphenyl thiourea, 1-2,4,6-trimethylphenyl-2-thiourea, 2,4,6-trimethyl-phenyl-thiourea, amino 2,4,6-trimethylphenyl amino methane-1-thione, 1-mesitylthiourea, acmc-20akda, maybridge1_006982 | |
(2,4,6-trimethylphenyl)thiourea |
Specifications
91059-97-1 | |
C10H14N2S | |
UN2811 | |
n-2,4,6-trimethylphenyl thiourea, n-mesitylthiourea, 2,4,6-trimethylphenyl thiourea, 1-2,4,6-trimethylphenyl thiourea, 1-2,4,6-trimethylphenyl-2-thiourea, 2,4,6-trimethyl-phenyl-thiourea, amino 2,4,6-trimethylphenyl amino methane-1-thione, 1-mesitylthiourea, acmc-20akda, maybridge1_006982 | |
CC1=CC(=C(C(=C1)C)NC(=S)N)C | |
194.296 | |
194.3 | |
N- (2,4,6-Trimethylphenyl)thiourea |
230°C to 231°C | |
MFCD00051488 | |
2642667 | |
LBUGQLLKHAHCJJ-UHFFFAOYSA-N | |
(2,4,6-trimethylphenyl)thiourea | |
2731118 | |
99% |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P264b-P270-P301+P310-P330-P501c
H301
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures