Learn More
N-[(2-Chloropyridin-4-yl)methyl]-N-methylamine, 97%, Thermo Scientific™
£81.33 - £569.00
Chemical Identifiers
CAS | 748187-76-0 |
---|---|
Molecular Formula | C7H9ClN2 |
Molecular Weight (g/mol) | 156.61 |
MDL Number | MFCD06654462 |
InChI Key | NIBLXLUVASMQFM-UHFFFAOYSA-N |
PubChem CID | 2794168 |
IUPAC Name | 1-(2-chloropyridin-4-yl)-N-methylmethanamine |
SMILES | CNCC1=CC(Cl)=NC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10184004
|
Thermo Scientific Maybridge
CC53646CB |
250 mg |
£81.33
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10165943
|
Thermo Scientific Maybridge
CC53646DA |
1 g |
£187.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10194004
|
Thermo Scientific Maybridge
CC53646DE |
5 g |
£569.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
748187-76-0 | |
156.61 | |
NIBLXLUVASMQFM-UHFFFAOYSA-N | |
1-(2-chloropyridin-4-yl)-N-methylmethanamine |
C7H9ClN2 | |
MFCD06654462 | |
2794168 | |
CNCC1=CC(Cl)=NC=C1 |
Specifications
748187-76-0 | |
100.0 | |
Amber glass bottle | |
MFCD06654462 | |
NIBLXLUVASMQFM-UHFFFAOYSA-N | |
1-(2-chloropyridin-4-yl)-N-methylmethanamine | |
2794168 | |
97% |
96.5 | |
97% | |
C7H9ClN2 | |
n-2-chloropyridin-4-yl methyl-n-methylamine, 2-chloropyridin-4-yl methyl methyl amine, 1-2-chloropyridin-4-yl-n-methylmethanamine, 2-chloro-4-methylamino methyl pyridine, 4-pyridinemethanamine,2-chloro-n-methyl, 2-chloro 4-pyridyl methyl methylamine, 4-pyridinemethanamine, 2-chloro-n-methyl, 2-chloro-n-methyl-4-pyridinemethanamine, 2-chloro-pyridin-4-ylmethyl-methyl-amine | |
CNCC1=CC(Cl)=NC=C1 | |
156.61 | |
156.62 | |
N-[(2-Chloropyridin-4-yl)methyl]-N-methylamine |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye damage.
Harmful if inhaled.
Causes severe skin burns and eye damage.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specifica
GHS Signal Word: Danger
Your input is important to us. Please complete this form to provide feedback related to the content on this product.