missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(2-Cyanoethyl)-N-(2-hydroxyethyl)aniline, 98%, Thermo Scientific™
£20.40 - £65.90
Chemical Identifiers
CAS | 92-64-8 |
---|---|
Molecular Formula | C11H14N2O |
Molecular Weight (g/mol) | 190.246 |
MDL Number | MFCD00035705 |
InChI Key | WZJJWQVBLSPALW-UHFFFAOYSA-N |
Synonym | n-cyanoethyl-n-hydroxyethylaniline, n-2-cyanoethyl-n-2-hydroxyethyl aniline, emery 5724, n-cyanoethyl-hydroxyethyl aniline, propanenitrile, 3-2-hydroxyethyl phenylamino, n-cyanoethyl-n-hydroxyethyl aniline, aniline, n-cyanoethyl-n-hydroxyethyl, 3-2-hydroxyethyl phenylamino propanenitrile, 3-n-2-hydroxyethyl anilino propiononitrile, propionitrile, 3-2-hydroxyethyl phenylamino |
PubChem CID | 113349 |
IUPAC Name | 3-[N-(2-hydroxyethyl)anilino]propanenitrile |
SMILES | C1=CC=C(C=C1)N(CCC#N)CCO |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsN-(2-Cyanoethyl)-N-(2-hydroxyethyl)aniline is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
Solubility
Not miscible or difficult to mix with water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with bases and oxidizing agents.
Chemical Identifiers
92-64-8 | |
190.246 | |
WZJJWQVBLSPALW-UHFFFAOYSA-N | |
113349 | |
C1=CC=C(C=C1)N(CCC#N)CCO |
C11H14N2O | |
MFCD00035705 | |
n-cyanoethyl-n-hydroxyethylaniline, n-2-cyanoethyl-n-2-hydroxyethyl aniline, emery 5724, n-cyanoethyl-hydroxyethyl aniline, propanenitrile, 3-2-hydroxyethyl phenylamino, n-cyanoethyl-n-hydroxyethyl aniline, aniline, n-cyanoethyl-n-hydroxyethyl, 3-2-hydroxyethyl phenylamino propanenitrile, 3-n-2-hydroxyethyl anilino propiononitrile, propionitrile, 3-2-hydroxyethyl phenylamino | |
3-[N-(2-hydroxyethyl)anilino]propanenitrile |
Specifications
92-64-8 | |
107°C to 110°C (15 mmHg) | |
1.5705 | |
2369849 | |
Not miscible or difficult to mix with water. | |
C1=CC=C(C=C1)N(CCC#N)CCO | |
190.246 | |
190.25 | |
N-(2-Cyanoethyl)-N-(2-hydroxyethyl)aniline |
1.12 | |
C11H14N2O | |
MFCD00035705 | |
n-cyanoethyl-n-hydroxyethylaniline, n-2-cyanoethyl-n-2-hydroxyethyl aniline, emery 5724, n-cyanoethyl-hydroxyethyl aniline, propanenitrile, 3-2-hydroxyethyl phenylamino, n-cyanoethyl-n-hydroxyethyl aniline, aniline, n-cyanoethyl-n-hydroxyethyl, 3-2-hydroxyethyl phenylamino propanenitrile, 3-n-2-hydroxyethyl anilino propiononitrile, propionitrile, 3-2-hydroxyethyl phenylamino | |
WZJJWQVBLSPALW-UHFFFAOYSA-N | |
3-[N-(2-hydroxyethyl)anilino]propanenitrile | |
113349 | |
98% |
Safety and Handling
missing translation for 'einecsNumber' : 202-174-5
missing translation for 'rtecsNumber' : UG2905000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures