missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Hydroxyethyl)ethylenediamine, 99%, Thermo Scientific Chemicals

Product Code. 11439792
Click to view available options
Quantity:
100 g
500 g
2500 g

It is used as an important intermediate in the manufacture of lube oil additives, fuel additives, chelating agents, surfactants and fabric softeners among other applications. N-(2-Hydroxyethyl)ethylenediamine [N-(2-Aminoethyl)ethanolamine] is used to study the aerobic biodecomposition of amines in hypersaline wastewaters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an important intermediate in the manufacture of lube oil additives, fuel additives, chelating agents, surfactants and fabric softeners among other applications. N-(2-Hydroxyethyl)ethylenediamine [N-(2-Aminoethyl)ethanolamine] is used to study the aerobic biodecomposition of amines in hypersaline wastewaters.

Solubility
Fully miscible in water.

Notes
Air Sensitive & Hygroscopic. Store under inert gas. Keep away from incompatibles.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-41-1
Molecular Formula C4H12N2O
Molecular Weight (g/mol) 104.15
MDL Number MFCD00008170
InChI Key LHIJANUOQQMGNT-UHFFFAOYSA-N
Synonym 2-2-aminoethylamino ethanol, n-2-hydroxyethyl ethylenediamine, aminoethylethanolamine, n-2-aminoethyl ethanolamine, ethanol, 2-2-aminoethyl amino, 2-2-aminoethyl amino ethanol, monoethanolethylenediamine, n-hydroxyethyl ethylenediamine, n-aminoethyl ethanolamine, 2-aminoethyl ethanolamine
PubChem CID 8112
IUPAC Name 2-(2-aminoethylamino)ethanol
SMILES NCCNCCO

Specifications

Melting Point -28°C
Density 1.029
pH 11.8
Boiling Point 238°C to 240°C
Flash Point 125°C (257°F)
Odor Amine-like
Refractive Index 1.4861
Quantity 2500 g
UN Number UN2735
Beilstein 506012
Sensitivity Air sensitive; Hygroscopic
Solubility Information Fully miscible in water.
Formula Weight 104.15
Percent Purity 99%
Chemical Name or Material N- (2-Hydroxyethyl)ethylenediamine
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.