missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Hydroxyethyl)piperazine, 98.5%, Thermo Scientific Chemicals

Product Code. 10498840
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-76-4
Molecular Formula C6H14N2O
Molecular Weight (g/mol) 130.19
MDL Number MFCD00005970
InChI Key WFCSWCVEJLETKA-UHFFFAOYSA-N
Synonym n-2-hydroxyethyl piperazine, 1-piperazineethanol, 1-2-hydroxyethyl piperazine, hydroxyethylpiperazine, 2-1-piperazinyl ethanol, 2-hydroxyethylpiperazine, 2-piperazinoethanol, 2-piperazin-1-yl ethan-1-ol, 1-piperazinethanol, 4-2-hydroxyethyl piperazine
PubChem CID 7677
IUPAC Name 2-piperazin-1-ylethanol
SMILES C1CN(CCN1)CCO

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point -38.5°C
Color Colorless to Yellow
Density 1.0600g/mL
Boiling Point 245°C to 246°C
Flash Point 135°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Refractive Index 1.5055 to 1.5075
Quantity 500 g
Beilstein 23, II, 6
Specific Gravity 1.06
Solubility Information Solubility in water: miscible with water. Other solubilities: miscible with most common organic solvents
Formula Weight 130.19
Percent Purity 98.50%
Physical Form Oily Liquid
Chemical Name or Material N-(2-Hydroxyethyl)piperazine, 98.5%
Show More Show Less
compliance-icons
Product Identifier
  • N-(2-Hydroxyethyl)piperazine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'