missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Hydroxyethyl)pyrrolidine, 97%, Thermo Scientific Chemicals

Product Code. 15446828
Click to view available options
Quantity:
25 mL
100 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs and is also used in medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs and is also used in medicine.

Solubility
Partly soluble in water. Soluble in alcohol, ether, benzene.

Notes
Store at room temperature. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2955-88-6
Molecular Formula C6H13NO
Molecular Weight (g/mol) 115.18
MDL Number MFCD00003181
InChI Key XBRDBODLCHKXHI-UHFFFAOYSA-N
Synonym 1-pyrrolidineethanol, n-2-hydroxyethyl pyrrolidine, 1-2-hydroxyethyl pyrrolidine, 2-pyrrolidin-1-yl ethanol, epolamine, 2-pyrrolidinoethanol, pyrrolidinoethanol, hydroxyethylpyrrolidine, 2-1-pyrrolidinyl ethanol, 2-pyrrolidin-1-yl ethan-1-ol
PubChem CID 76288
ChEBI CHEBI:48293
IUPAC Name 2-pyrrolidin-1-ylethanol
SMILES OCCN1CCCC1

Specifications

Density 0.985
Boiling Point 79°C to 81°C
Flash Point 56°C (133°F)
Refractive Index 1.471
Quantity 25 mL
UN Number UN1993
Beilstein 102976
Solubility Information Partly soluble in water. Soluble in alcohol,ether,benzene.
Formula Weight 115.17
Percent Purity 97%
Chemical Name or Material N-(2-Hydroxyethyl)pyrrolidine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'