missing translation for 'onlineSavingsMsg'
Learn More

N-[2-(Trifluoromethyl)phenyl]thiourea, 98%, Thermo Scientific™

Product Code. 11307396
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1736-71-6
Molecular Formula C8H7F3N2S
Molecular Weight (g/mol) 220.213
MDL Number MFCD00041161
InChI Key FVXFFFHGYOYYQX-UHFFFAOYSA-N
Synonym 2-trifluoromethyl phenylthiourea, 1-2-trifluoromethyl phenyl-2-thiourea, 2-trifluoromethyl phenyl thiourea, 1-2-trifluoromethyl phenyl thiourea, 2-trifluoromethyl-phenyl-thiourea, thiourea, 2-trifluoromethyl phenyl, thiourea, n-2-trifluoromethyl phenyl
PubChem CID 737227
IUPAC Name [2-(trifluoromethyl)phenyl]thiourea
SMILES C1=CC=C(C(=C1)C(F)(F)F)NC(=S)N

Specifications

Melting Point 163°C to 165°C
Odor Odorless
Quantity 5 g
UN Number UN2811
Beilstein 2695476
Formula Weight 220.22
Percent Purity 98%
Chemical Name or Material N-[2-(Trifluoromethyl)phenyl]thiourea
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'