missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-[2-(Trifluoromethyl)phenyl]thiourea, 98%, Thermo Scientific™
£23.90 - £79.00
Chemical Identifiers
CAS | 1736-71-6 |
---|---|
Molecular Formula | C8H7F3N2S |
Molecular Weight (g/mol) | 220.213 |
MDL Number | MFCD00041161 |
InChI Key | FVXFFFHGYOYYQX-UHFFFAOYSA-N |
Synonym | 2-trifluoromethyl phenylthiourea, 1-2-trifluoromethyl phenyl-2-thiourea, 2-trifluoromethyl phenyl thiourea, 1-2-trifluoromethyl phenyl thiourea, 2-trifluoromethyl-phenyl-thiourea, thiourea, 2-trifluoromethyl phenyl, thiourea, n-2-trifluoromethyl phenyl |
PubChem CID | 737227 |
IUPAC Name | [2-(trifluoromethyl)phenyl]thiourea |
SMILES | C1=CC=C(C(=C1)C(F)(F)F)NC(=S)N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11397386
|
Thermo Scientific Alfa Aesar
L09377.03 |
1 g |
£23.90
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11307396
|
Thermo Scientific Alfa Aesar
L09377.06 |
5 g |
£79.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
1736-71-6 | |
220.213 | |
FVXFFFHGYOYYQX-UHFFFAOYSA-N | |
737227 | |
C1=CC=C(C(=C1)C(F)(F)F)NC(=S)N |
C8H7F3N2S | |
MFCD00041161 | |
2-trifluoromethyl phenylthiourea, 1-2-trifluoromethyl phenyl-2-thiourea, 2-trifluoromethyl phenyl thiourea, 1-2-trifluoromethyl phenyl thiourea, 2-trifluoromethyl-phenyl-thiourea, thiourea, 2-trifluoromethyl phenyl, thiourea, n-2-trifluoromethyl phenyl | |
[2-(trifluoromethyl)phenyl]thiourea |
Specifications
1736-71-6 | |
Odorless | |
MFCD00041161 | |
2695476 | |
FVXFFFHGYOYYQX-UHFFFAOYSA-N | |
[2-(trifluoromethyl)phenyl]thiourea | |
737227 | |
98% |
163°C to 165°C | |
C8H7F3N2S | |
UN2811 | |
2-trifluoromethyl phenylthiourea, 1-2-trifluoromethyl phenyl-2-thiourea, 2-trifluoromethyl phenyl thiourea, 1-2-trifluoromethyl phenyl thiourea, 2-trifluoromethyl-phenyl-thiourea, thiourea, 2-trifluoromethyl phenyl, thiourea, n-2-trifluoromethyl phenyl | |
C1=CC=C(C(=C1)C(F)(F)F)NC(=S)N | |
220.213 | |
220.22 | |
N-[2-(Trifluoromethyl)phenyl]thiourea |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P264b-P270-P301+P310-P330-P501c
H301
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 217-084-1
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures