missing translation for 'onlineSavingsMsg'
Learn More

N-[3-(2-Furyl)benzyl]-N-methylamine, ≥97%, Thermo Scientific™

Product Code. 10225844
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 857284-27-6
Molecular Formula C12H13NO
Molecular Weight (g/mol) 187.242
MDL Number MFCD08435877
InChI Key WMCXJYGZLCREMM-UHFFFAOYSA-N
Synonym n-3-2-furyl benzyl-n-methylamine, 3-furan-2-yl phenyl methyl methyl amine, 1-3-furan-2-yl phenyl-n-methylmethanamine, 3-2-furyl phenyl methyl methylamine, benzenemethanamine,3-2-furanyl-n-methyl
PubChem CID 18525790
IUPAC Name 1-[3-(furan-2-yl)phenyl]-N-methylmethanamine
SMILES CNCC1=CC=CC(=C1)C2=CC=CO2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 1 g
Formula Weight 187.24
Percent Purity 97%
Chemical Name or Material N-[3-(2-furyl)benzyl]-N-methylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'