missing translation for 'onlineSavingsMsg'
Learn More

N-(3,5-Dichlorophenyl)thiourea, 98+%, Thermo Scientific™

Product Code. 11467877
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107707-33-5
Molecular Formula C7H6Cl2N2S
Molecular Weight (g/mol) 221.099
MDL Number MFCD00041168
InChI Key BNJARXOEVLYKDN-UHFFFAOYSA-N
Synonym 1-3,5-dichlorophenyl thiourea, n-3,5-dichlorophenyl thiourea, 3,5-dichlorophenyl thiourea, 1-3,5-dichlorophenyl-2-thiourea, thiourea, 3,5-dichlorophenyl, amino 3,5-dichlorophenyl amino methane-1-thione, thiourea;, maybridge1_007644, acmc-1bnq2
PubChem CID 2737539
IUPAC Name (3,5-dichlorophenyl)thiourea
SMILES C1=C(C=C(C=C1Cl)Cl)NC(=S)N

Specifications

Melting Point 175°C to 179°C
Quantity 5 g
UN Number UN2811
Beilstein 3257049
Formula Weight 221.11
Percent Purity ≥98%
Chemical Name or Material N-(3,5-Dichlorophenyl)thiourea
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'