missing translation for 'onlineSavingsMsg'
Learn More

N-(3-Acetyl-4-hydroxyphenyl)butyramide, 97%, Thermo Scientific Chemicals

£24.30 - £258.00

Chemical Identifiers

CAS 40188-45-2
Molecular Formula C12H15NO3
Molecular Weight (g/mol) 221.26
MDL Number MFCD00798556
InChI Key FGWZEOPEZISTTR-UHFFFAOYSA-N
Synonym 2-acetyl-4-butyramidophenol, n-3-acetyl-4-hydroxyphenyl butanamide, n-3-acetyl-4-hydroxyphenyl butyramide, ac-hophe-oh, unii-14qjb65r4v, 3'-acetyl-4'-hydroxybutyranilide, butanamide, n-3-acetyl-4-hydroxyphenyl, 5'-butyramido-2'-hydroxyacetophenone, acebutolol impurity c, n-3-acetyl-4-hydroxyphenyl butanamid
PubChem CID 736331
IUPAC Name N-(3-acetyl-4-hydroxyphenyl)butanamide
SMILES CCCC(=O)NC1=CC=C(O)C(=C1)C(C)=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11482958
View Documents
Thermo Scientific Alfa Aesar
A18672.18
50 g
£24.30
50g
Please to purchase this item. Need a web account? Register with us today!
11492958
View Documents
Thermo Scientific Alfa Aesar
A18672.30
250 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11472958
View Documents
Thermo Scientific Alfa Aesar
A18672.0B
1000 g
£258.00
1000g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

40188-45-2
221.26
FGWZEOPEZISTTR-UHFFFAOYSA-N
736331
CCCC(=O)NC1=CC=C(O)C(=C1)C(C)=O
C12H15NO3
MFCD00798556
2-acetyl-4-butyramidophenol, n-3-acetyl-4-hydroxyphenyl butanamide, n-3-acetyl-4-hydroxyphenyl butyramide, ac-hophe-oh, unii-14qjb65r4v, 3'-acetyl-4'-hydroxybutyranilide, butanamide, n-3-acetyl-4-hydroxyphenyl, 5'-butyramido-2'-hydroxyacetophenone, acebutolol impurity c, n-3-acetyl-4-hydroxyphenyl butanamid
N-(3-acetyl-4-hydroxyphenyl)butanamide

Specifications

40188-45-2
C12H15NO3
2-acetyl-4-butyramidophenol, n-3-acetyl-4-hydroxyphenyl butanamide, n-3-acetyl-4-hydroxyphenyl butyramide, ac-hophe-oh, unii-14qjb65r4v, 3'-acetyl-4'-hydroxybutyranilide, butanamide, n-3-acetyl-4-hydroxyphenyl, 5'-butyramido-2'-hydroxyacetophenone, acebutolol impurity c, n-3-acetyl-4-hydroxyphenyl butanamid
CCCC(=O)NC1=CC=C(O)C(=C1)C(C)=O
221.26
221.26
N-(3-Acetyl-4-hydroxyphenyl)butyramide
124°C to 126°C
MFCD00798556
FGWZEOPEZISTTR-UHFFFAOYSA-N
N-(3-acetyl-4-hydroxyphenyl)butanamide
736331
97%
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 254-831-0

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only