missing translation for 'onlineSavingsMsg'
Learn More
N-(3-Formyl-4-pyridinyl)-2,2-dimethylpropanamide, 97%, Thermo Scientific™ Product Code.: 10703823

N-(3-Formyl-4-pyridinyl)-2,2-dimethylpropanamide, 97%, Thermo Scientific™

Product Code. 10703823
250 mg, Amber Glass Bottle
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 10703823

Brand: Thermo Scientific Maybridge MO00095CB

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 86847-71-4
Molecular Formula C11H14N2O2
Molecular Weight (g/mol) 206.245
MDL Number MFCD03086208
InChI Key ICMXCEJBHWHTBH-UHFFFAOYSA-N
Synonym n-3-formylpyridin-4-yl pivalamide, n-3-formyl-4-pyridinyl-2,2-dimethylpropanamide, n-3-formylpyridin-4-yl-2,2-dimethylpropanamide, n-3-formyl-pyridin-4-yl-2,2-dimethyl-propionamide, 4-2,2-dimethylpropanoyl amino pyridine-3-carboxaldehyde, 2,2-dimethyl-n-3-formylpyridin-4-yl propanamide, 4-2,2-dimethylpropanoyl amino nicotinaldehyde, propanamide,n-3-formyl-4-pyridinyl-2,2-dimethyl, 4-2,2-dimethylpropanoyl amino-3-formylpyridine, n-3-formylpyrdn-4-yl pvalamde
PubChem CID 2779664
IUPAC Name N-(3-formylpyridin-4-yl)-2,2-dimethylpropanamide
SMILES CC(C)(C)C(=O)NC1=C(C=NC=C1)C=O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 206.24
Percent Purity 97%
Chemical Name or Material N-(3-Formyl-4-pyridinyl)-2, 2-dimethylpropanamide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N-(3-Formyl-4-pyridinyl)-2,2-dimethylpropanamide, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.