missing translation for 'onlineSavingsMsg'
Learn More

N-(3-Hydroxyphenyl)thiourea, 97%

Product Code. 11302387
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
This item is not returnable. View return policy

Product Code. 11302387

Brand: Thermo Scientific Alfa Aesar L09975.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3394-05-6 | C8H8N2O6 | 228.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3394-05-6
MDL Number MFCD00022167

Specifications

Melting Point 175°C to 180°C
Odor Odorless
Molecular Formula C7H8N2OS
Quantity 10 g
Beilstein 2090372
Synonym 1-3-hydroxyphenyl-2-thiourea, 3-hydroxyphenyl thiourea, 1-3-hydroxyphenyl thiourea, 3-aminothioxomethyl amino phenol, acmc-20apbj, 1-m-hydroxyphenyl thiourea, 3-hydroxy-phenylthioharnstoff, thiourea, 3-hydroxyphenyl, thiourea,n-3-hydroxyphenyl, 1-m-hydroxyphenyl-2-thiourea
InChI Key BHJYKFUCQNISJA-UHFFFAOYSA-N
SMILES C1=CC(=CC(=C1)O)NC(=S)N
IUPAC Name (3-hydroxyphenyl)thiourea
Molecular Weight (g/mol) 168.21
PubChem CID 2759330
Formula Weight 168.22
Percent Purity 97%
Chemical Name or Material N-(3-Hydroxyphenyl)thiourea
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.