missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Bromo-3-methylphenyl)-2-methoxybenzamide, 97%, Thermo Scientific™

Product Code. 15465186
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 586394-64-1
Molecular Formula C15H14BrNO2
Molecular Weight (g/mol) 320.186
MDL Number MFCD03241922
InChI Key DAJGBNVYZLWLSV-UHFFFAOYSA-N
Synonym n-4-bromo-3-methylphenyl-2-methoxybenzamide, cambridge id 5937162
PubChem CID 2877851
IUPAC Name N-(4-bromo-3-methylphenyl)-2-methoxybenzamide
SMILES CC1=C(C=CC(=C1)NC(=O)C2=CC=CC=C2OC)Br

Specifications

Quantity 250 mg
Formula Weight 320.19
Percent Purity 97%
Chemical Name or Material N-(4-Bromo-3-methylphenyl)-2-methoxybenzamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'