missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Bromophenyl)maleamic acid, 97%, Thermo Scientific™

Product Code. 11410708
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36847-86-6
Molecular Formula C10H8BrNO3
Molecular Weight (g/mol) 270.082
MDL Number MFCD00134952
InChI Key DSNFUZCLEZJCRN-WAYWQWQTSA-N
Synonym n-4-bromophenyl maleamic acid, 4-4-bromoanilino-4-oxobut-2-enoic acid, 4-4-bromophenyl amino-4-oxobut-2-enoic acid, 2z-3-4-bromophenyl carbamoyl prop-2-enoic acid, 3-4-bromophenyl carbamoyl prop-2-enoic acid, n-4-bromophenyl maleamidic acid, 3-4-bromo-phenylcarbamoyl-acrylic acid, z-4-4-bromoanilino-4-oxobut-2-enoic acid, 2-butenoic acid, 4-4-bromophenyl amino-4-oxo-, 2z
PubChem CID 1551067
IUPAC Name (Z)-4-(4-bromoanilino)-4-oxobut-2-enoic acid
SMILES C1=CC(=CC=C1NC(=O)C=CC(=O)O)Br

Specifications

Quantity 5 g
Formula Weight 270.08
Percent Purity 97%
Chemical Name or Material N-(4-Bromophenyl)maleamic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'