missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(4-Bromophenyl)maleamic acid, 97%, Thermo Scientific™
£20.59 - £312.00
Chemical Identifiers
CAS | 36847-86-6 |
---|---|
Molecular Formula | C10H8BrNO3 |
Molecular Weight (g/mol) | 270.082 |
MDL Number | MFCD00134952 |
InChI Key | DSNFUZCLEZJCRN-WAYWQWQTSA-N |
Synonym | n-4-bromophenyl maleamic acid, 4-4-bromoanilino-4-oxobut-2-enoic acid, 4-4-bromophenyl amino-4-oxobut-2-enoic acid, 2z-3-4-bromophenyl carbamoyl prop-2-enoic acid, 3-4-bromophenyl carbamoyl prop-2-enoic acid, n-4-bromophenyl maleamidic acid, 3-4-bromo-phenylcarbamoyl-acrylic acid, z-4-4-bromoanilino-4-oxobut-2-enoic acid, 2-butenoic acid, 4-4-bromophenyl amino-4-oxo-, 2z |
PubChem CID | 1551067 |
IUPAC Name | (Z)-4-(4-bromoanilino)-4-oxobut-2-enoic acid |
SMILES | C1=CC(=CC=C1NC(=O)C=CC(=O)O)Br |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
36847-86-6 | |
270.082 | |
DSNFUZCLEZJCRN-WAYWQWQTSA-N | |
1551067 | |
C1=CC(=CC=C1NC(=O)C=CC(=O)O)Br |
C10H8BrNO3 | |
MFCD00134952 | |
n-4-bromophenyl maleamic acid, 4-4-bromoanilino-4-oxobut-2-enoic acid, 4-4-bromophenyl amino-4-oxobut-2-enoic acid, 2z-3-4-bromophenyl carbamoyl prop-2-enoic acid, 3-4-bromophenyl carbamoyl prop-2-enoic acid, n-4-bromophenyl maleamidic acid, 3-4-bromo-phenylcarbamoyl-acrylic acid, z-4-4-bromoanilino-4-oxobut-2-enoic acid, 2-butenoic acid, 4-4-bromophenyl amino-4-oxo-, 2z | |
(Z)-4-(4-bromoanilino)-4-oxobut-2-enoic acid |
Specifications
36847-86-6 | |
MFCD00134952 | |
DSNFUZCLEZJCRN-WAYWQWQTSA-N | |
(Z)-4-(4-bromoanilino)-4-oxobut-2-enoic acid | |
1551067 | |
97% |
C10H8BrNO3 | |
n-4-bromophenyl maleamic acid, 4-4-bromoanilino-4-oxobut-2-enoic acid, 4-4-bromophenyl amino-4-oxobut-2-enoic acid, 2z-3-4-bromophenyl carbamoyl prop-2-enoic acid, 3-4-bromophenyl carbamoyl prop-2-enoic acid, n-4-bromophenyl maleamidic acid, 3-4-bromo-phenylcarbamoyl-acrylic acid, z-4-4-bromoanilino-4-oxobut-2-enoic acid, 2-butenoic acid, 4-4-bromophenyl amino-4-oxo-, 2z | |
C1=CC(=CC=C1NC(=O)C=CC(=O)O)Br | |
270.082 | |
270.08 | |
N-(4-Bromophenyl)maleamic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures