missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Chloro-2-butynyl)phthalimide, 97%, Thermo Scientific Chemicals

Product Code. 11312987
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

N-(4-Chloro-2-butynyl)phthalimide is used to make other chemicals and as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-(4-Chloro-2-butynyl)phthalimide is used to make other chemicals and as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong bases and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4819-69-6
Molecular Formula C12H8ClNO2
Molecular Weight (g/mol) 233.651
MDL Number MFCD01318120
InChI Key TXNDRPKNOXQAAO-UHFFFAOYSA-N
Synonym n-4-chloro-2-butynyl phthalimide, 2-4-chlorobut-2-yn-1-yl isoindoline-1,3-dione, 2-4-chlorobut-2-yn-1-yl-1h-isoindole-1,3 2h-dione, 2-4-chlorobut-2-yn-1-yl isoindole-1,3-dione, acmc-20akbi, n-4-chlorobut-2-ynyl phthalimide, 2-4-chlorobut-2-ynyl isoindole-1,3-dione, 2-4-chloro-2-butynyl isoindoline-1,3-dione, 1h-isoindole-1,3 2h-dione, 2-4-chloro-2-butyn-1-yl, 2-4-chlorobut-2-yn-1-yl-2,3-dihydro-1h-isoindole-1,3-dione
PubChem CID 237537
IUPAC Name 2-(4-chlorobut-2-ynyl)isoindole-1,3-dione
SMILES C1=CC=C2C(=C1)C(=O)N(C2=O)CC#CCCl

Specifications

Melting Point 115°C to 120°C
Odor Nearly Odorless
Quantity 1 g
UN Number UN3261
Beilstein 179626
Solubility Information Insoluble in water.
Formula Weight 233.66
Percent Purity 97%
Chemical Name or Material N-(4-Chloro-2-butynyl)phthalimide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'