missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Fluorobenzyl)-2-(trifluoromethyl)aniline, 97%, Thermo Scientific™

Product Code. 15442866
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1041583-33-8
Molecular Formula C14H11F4N
Molecular Weight (g/mol) 269.24
MDL Number MFCD12618204
InChI Key JCWUQELJSDRKEP-UHFFFAOYSA-N
Synonym n-4-fluorobenzyl-2-trifluoromethyl aniline, n-4-fluorophenyl methyl-2-trifluoromethyl aniline
PubChem CID 29292080
IUPAC Name N-[(4-fluorophenyl)methyl]-2-(trifluoromethyl)aniline
SMILES FC1=CC=C(CNC2=CC=CC=C2C(F)(F)F)C=C1

Specifications

Quantity 250 mg
Formula Weight 269.24
Percent Purity 97%
Chemical Name or Material N-(4-Fluorobenzyl)-2-(trifluoromethyl)aniline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'