missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(4-Hydroxyphenyl)thiourea, 98+%, Thermo Scientific Chemicals
£26.10 - £80.20
Chemical Identifiers
CAS | 1520-27-0 |
---|---|
Molecular Formula | C7H8N2OS |
Molecular Weight (g/mol) | 168.214 |
MDL Number | MFCD00041186 |
InChI Key | QICKOOCQSYZYQB-UHFFFAOYSA-N |
Synonym | 1-4-hydroxyphenyl-2-thiourea, 1-4-hydroxyphenyl thiourea, 4-hydroxyphenyl thiourea, p-hydroxyphenylthiourea, usaf b-75, n-4-hydroxyphenyl thiourea, urea, 1-p-hydroxyphenyl-2-thio, thiourea, 4-hydroxyphenyl, 4-hydroxy-phenyl-thiourea, 4-aminothioxomethyl amino phenol |
PubChem CID | 2759331 |
IUPAC Name | (4-hydroxyphenyl)thiourea |
SMILES | C1=CC(=CC=C1NC(=S)N)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11333226
|
Thermo Scientific Alfa Aesar
L12537.03 |
1 g |
£26.10
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11343226
|
Thermo Scientific Alfa Aesar
L12537.06 |
5 g |
£80.20
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1520-27-0 | |
168.214 | |
QICKOOCQSYZYQB-UHFFFAOYSA-N | |
2759331 | |
C1=CC(=CC=C1NC(=S)N)O |
C7H8N2OS | |
MFCD00041186 | |
1-4-hydroxyphenyl-2-thiourea, 1-4-hydroxyphenyl thiourea, 4-hydroxyphenyl thiourea, p-hydroxyphenylthiourea, usaf b-75, n-4-hydroxyphenyl thiourea, urea, 1-p-hydroxyphenyl-2-thio, thiourea, 4-hydroxyphenyl, 4-hydroxy-phenyl-thiourea, 4-aminothioxomethyl amino phenol | |
(4-hydroxyphenyl)thiourea |
Specifications
1520-27-0 | |
Odorless | |
MFCD00041186 | |
1-4-hydroxyphenyl-2-thiourea, 1-4-hydroxyphenyl thiourea, 4-hydroxyphenyl thiourea, p-hydroxyphenylthiourea, usaf b-75, n-4-hydroxyphenyl thiourea, urea, 1-p-hydroxyphenyl-2-thio, thiourea, 4-hydroxyphenyl, 4-hydroxy-phenyl-thiourea, 4-aminothioxomethyl amino phenol | |
C1=CC(=CC=C1NC(=S)N)O | |
168.214 | |
168.22 | |
N-(4-Hydroxyphenyl)thiourea |
∽210°C (decomposition) | |
C7H8N2OS | |
1102429 | |
QICKOOCQSYZYQB-UHFFFAOYSA-N | |
(4-hydroxyphenyl)thiourea | |
2759331 | |
≥98% |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
missing translation for 'rtecsNumber' : YT5250000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only