missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Methoxyphenyl)-3-nitrobenzenesulfonamide, 97%, Thermo Scientific™

Product Code. 15414006
Click to view available options
Quantity:
100 mg
500 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 180631-81-6
Molecular Formula C13H12N2O5S
Molecular Weight (g/mol) 308.31
MDL Number MFCD00414861
InChI Key NHEAQAMNXXRBQT-UHFFFAOYSA-N
Synonym n-4-methoxyphenyl-3-nitrobenzenesulfonamide, cambridge id 5135326, 4-methoxyphenyl 3-nitrophenyl sulfonyl amine, n-4-methoxy-phenyl-3-nitro-benzenesulfonamide
PubChem CID 2829315
IUPAC Name N-(4-methoxyphenyl)-3-nitrobenzenesulfonamide
SMILES COC1=CC=C(NS(=O)(=O)C2=CC=CC(=C2)[N+]([O-])=O)C=C1

Specifications

Color Pink
Quantity 100 mg
Beilstein 2764039
Formula Weight 308.3
Percent Purity 97%
Physical Form Solid
Chemical Name or Material N-(4-Methoxyphenyl)-3-nitrobenzenesulfonamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'