missing translation for 'onlineSavingsMsg'
Learn More
N-(4-Piperidin-1-ylbenzyl)cyclopropanamine, ≥95%, Thermo Scientific™ Product Code.: 10331063

N-(4-Piperidin-1-ylbenzyl)cyclopropanamine, ≥95%, Thermo Scientific™

Product Code. 10331063
250 mg, Amber Glass Bottle
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 10331063

Brand: Thermo Scientific Maybridge CC33958CB

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 857283-76-2
Molecular Formula C15H22N2
Molecular Weight (g/mol) 230.36
MDL Number MFCD07772814
InChI Key UNFZQBXRXGLJLF-UHFFFAOYSA-N
Synonym n-4-piperidin-1-ylbenzyl cyclopropanamine, n-4-piperidin-1-yl phenyl methyl cyclopropanamine, cyclopropyl 4-piperidylphenyl methyl amine, n-4-piperidin-1-yl benzyl cyclopropanamine, benzenemethanamine,n-cyclopropyl-4-1-piperidinyl, n-4-piperidin-1-ylphenyl methyl cyclopropanamine
PubChem CID 7162339
IUPAC Name N-{[4-(piperidin-1-yl)phenyl]methyl}cyclopropanamine
SMILES C(NC1CC1)C1=CC=C(C=C1)N1CCCCC1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 230.35
Percent Purity 95%
Chemical Name or Material N-(4-piperidin-1-ylbenzyl)cyclopropanamine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N-(4-Piperidin-1-ylbenzyl)cyclopropanamine, ≥95%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.