Learn More
N-(4-Pyridyl)thiourea, 97%, Thermo Scientific™
£38.00 - £208.00
Chemical Identifiers
CAS | 164670-44-4 |
---|---|
Molecular Formula | C6H7N3S |
Molecular Weight (g/mol) | 153.203 |
MDL Number | MFCD00068177 |
InChI Key | HLOFIQOOOSRNFY-UHFFFAOYSA-N |
Synonym | 4-pyridylthiourea, n-4-pyridyl thiourea, pyridin-4-yl thiourea, 1-pyridin-4-yl thiourea, 1-4-pyridyl-2-thiourea, pyridin-4-yl-thiourea, amino 4-pyridylamino methane-1-thione, acmc-20amay, n-pyridin-4-ylthiourea, 1-pyridin-4-ylthiourea |
PubChem CID | 2737249 |
IUPAC Name | pyridin-4-ylthiourea |
SMILES | C1=CN=CC=C1NC(=S)N |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10662263
|
Thermo Scientific Maybridge
SEW02754DA |
1 g | Amber Glass Bottle |
£38.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10532083
|
Thermo Scientific Maybridge
SEW02754EA |
10 g | Amber Glass Bottle |
£208.00
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
164670-44-4 | |
153.203 | |
HLOFIQOOOSRNFY-UHFFFAOYSA-N | |
2737249 | |
C1=CN=CC=C1NC(=S)N |
C6H7N3S | |
MFCD00068177 | |
4-pyridylthiourea, n-4-pyridyl thiourea, pyridin-4-yl thiourea, 1-pyridin-4-yl thiourea, 1-4-pyridyl-2-thiourea, pyridin-4-yl-thiourea, amino 4-pyridylamino methane-1-thione, acmc-20amay, n-pyridin-4-ylthiourea, 1-pyridin-4-ylthiourea | |
pyridin-4-ylthiourea |
Specifications
164670-44-4 | |
97% | |
MFCD00068177 | |
HLOFIQOOOSRNFY-UHFFFAOYSA-N | |
pyridin-4-ylthiourea | |
2737249 | |
97% |
97.0 | |
C6H7N3S | |
4-pyridylthiourea, n-4-pyridyl thiourea, pyridin-4-yl thiourea, 1-pyridin-4-yl thiourea, 1-4-pyridyl-2-thiourea, pyridin-4-yl-thiourea, amino 4-pyridylamino methane-1-thione, acmc-20amay, n-pyridin-4-ylthiourea, 1-pyridin-4-ylthiourea | |
C1=CN=CC=C1NC(=S)N | |
153.203 | |
153.21 | |
N-(4-Pyridyl)thiourea |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning