missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Pyridylmethyl)ethylamine, 96%, Thermo Scientific Chemicals

Product Code. 11303207
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33403-97-3
Molecular Formula C8H12N2
Molecular Weight (g/mol) 136.198
MDL Number MFCD00023632
InChI Key ZBAMQLFFVBPAOX-UHFFFAOYSA-N
Synonym 4-ethylaminomethyl pyridine, n-pyridin-4-ylmethyl ethanamine, n-4-pyridylmethyl ethylamine, 4-pyridinemethanamine, n-ethyl, unii-eye32x3w6x, n-ethyl-4-pyridinemethanamine, ethyl pyridin-4-ylmethyl amine, 4-ethylamino methyl pyridine, n-ethyl-4-picolylamine, eye32x3w6x
PubChem CID 96681
IUPAC Name N-(pyridin-4-ylmethyl)ethanamine
SMILES CCNCC1=CC=NC=C1

Specifications

Density 0.98
Boiling Point 103°C to 105°C (8 mmHg)
Flash Point 89°C (192°F)
Refractive Index 1.5175
Quantity 5 g
Beilstein 471685
Formula Weight 136.2
Percent Purity 96%
Chemical Name or Material N-(4-Pyridylmethyl)ethylamine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'