missing translation for 'onlineSavingsMsg'
Learn More

N-(4-tert-butylbenzyl)-N-methylamine, Thermo Scientific™

Product Code. p-4391241
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10321530

Brand: Thermo Scientific Maybridge MO08598DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 65542-26-9
Molecular Formula C12H19N
Molecular Weight (g/mol) 177.291
InChI Key ZSHCHOYJMLEAOX-UHFFFAOYSA-N
Synonym n-4-tert-butylbenzyl-n-methylamine, 4-tert-butylphenyl methyl methyl amine, 1-4-tert-butyl phenyl-n-methylmethanamine, 1-4-tert-butylphenyl-n-methylmethanamine, 4-tert-butylbenzylmethylamine, n-methyl-4-tert-butylbenzylamine, n-methyl-4-tert.butylbenzylamine, n-methyl 4-tert-butylbenzyl amine, 4-tert-butyl phenyl methyl methylamine, benzenemethanamine,4-1,1-dimethylethyl-n-methyl
PubChem CID 2396874
IUPAC Name 1-(4-tert-butylphenyl)-N-methylmethanamine
SMILES CC(C)(C)C1=CC=C(C=C1)CNC

Specifications

Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 177.29
Percent Purity 97%
Chemical Name or Material N-(4-tert-butylbenzyl)-N-methylamine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.