missing translation for 'onlineSavingsMsg'
Learn More

N-[4-(Trifluoromethyl)phenyl]thiourea, 98%, Thermo Scientific Chemicals

Product Code. 11308765
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1736-72-7
Molecular Formula C8H7F3N2S
Molecular Weight (g/mol) 220.21
MDL Number MFCD00041189
InChI Key OWTDDZMFRLUBQI-UHFFFAOYSA-N
Synonym 1-4-trifluoromethyl phenyl thiourea, 4-trifluoromethyl phenyl thiourea, 1-4-trifluoromethyl phenyl-2-thiourea, 4-trifluoromethyl phenylthiourea, n-4-trifluoromethyl phenyl thiourea, 4-trifluoromethyl-phenyl-thiourea, thiourea, 4-trifluoromethyl phenyl, amino 4-trifluoromethyl phenyl amino methane-1-thione
PubChem CID 2777727
IUPAC Name [4-(trifluoromethyl)phenyl]thiourea
SMILES NC(=S)NC1=CC=C(C=C1)C(F)(F)F

Specifications

Melting Point 142°C to 146°C
Odor Odorless
Quantity 5 g
UN Number UN2811
Beilstein 2695479
Formula Weight 220.22
Percent Purity 98%
Chemical Name or Material N-[4-(Trifluoromethyl)phenyl]thiourea

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'