missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(5-Fluoro-2-methylphenyl)thiourea, 97%, Thermo Scientific™
£16.70 - £241.00
Chemical Identifiers
CAS | 16822-86-9 |
---|---|
Molecular Formula | C8H9FN2S |
Molecular Weight (g/mol) | 184.23 |
MDL Number | MFCD00052380 |
InChI Key | NZTMKTPFNGHTJG-UHFFFAOYSA-N |
Synonym | 5-fluoro-2-methylphenyl thiourea, 1-5-fluoro-2-methylphenyl thiourea, 1-5-fluoro-2-methylphenyl-2-thiourea, 5-fluoro-2-methyl-phenyl thiourea, n-5-fluoro-2-methylphenyl thiourea, acmc-1c5tk, 5-fluoro-2-methyl-phenyl-thiourea, 1-5-fluoranyl-2-methyl-phenyl thiourea |
PubChem CID | 2732698 |
IUPAC Name | (5-fluoro-2-methylphenyl)thiourea |
SMILES | CC1=CC=C(F)C=C1NC(N)=S |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
16822-86-9 | |
184.23 | |
NZTMKTPFNGHTJG-UHFFFAOYSA-N | |
2732698 | |
CC1=CC=C(F)C=C1NC(N)=S |
C8H9FN2S | |
MFCD00052380 | |
5-fluoro-2-methylphenyl thiourea, 1-5-fluoro-2-methylphenyl thiourea, 1-5-fluoro-2-methylphenyl-2-thiourea, 5-fluoro-2-methyl-phenyl thiourea, n-5-fluoro-2-methylphenyl thiourea, acmc-1c5tk, 5-fluoro-2-methyl-phenyl-thiourea, 1-5-fluoranyl-2-methyl-phenyl thiourea | |
(5-fluoro-2-methylphenyl)thiourea |
Specifications
16822-86-9 | |
Odorless | |
MFCD00052380 | |
5-fluoro-2-methylphenyl thiourea, 1-5-fluoro-2-methylphenyl thiourea, 1-5-fluoro-2-methylphenyl-2-thiourea, 5-fluoro-2-methyl-phenyl thiourea, n-5-fluoro-2-methylphenyl thiourea, acmc-1c5tk, 5-fluoro-2-methyl-phenyl-thiourea, 1-5-fluoranyl-2-methyl-phenyl thiourea | |
CC1=CC=C(F)C=C1NC(N)=S | |
184.23 | |
184.23 | |
N-(5-Fluoro-2-methylphenyl)thiourea |
117°C to 120°C | |
C8H9FN2S | |
UN2811 | |
NZTMKTPFNGHTJG-UHFFFAOYSA-N | |
(5-fluoro-2-methylphenyl)thiourea | |
2732698 | |
97% |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P264b-P270-P301+P310-P330-P501c
H301
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures