missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(6-Bromohexyl)phthalimide, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
N-(6-Bromohexyl)phthalimide is used as a pharmaceutical intermediate and in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-(6-Bromohexyl)phthalimide is used as a pharmaceutical intermediate and in organic synthesis.
Solubility
Insoluble in water.
Notes
Incompatible with oxidizing agents.
N-(6-Bromohexyl)phthalimide is used as a pharmaceutical intermediate and in organic synthesis.
Solubility
Insoluble in water.
Notes
Incompatible with oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 54°C to 60°C |
Boiling Point | 186°C to 188°C (0.4 mmHg) |
Odor | Odorless |
Quantity | 25 g |
Beilstein | 186875 |
Synonym | n-6-bromohexyl phthalimide, 2-6-bromohexyl isoindoline-1,3-dione, n-6-bromohexyl-phthalimide, 2-6-bromohexyl-1h-isoindole-1,3 2h-dione, 2-6-bromohexyl isoindole-1,3-dione, 2-6-bromohexyl-2,3-dihydro-1h-isoindole-1,3-dione, 2-6-bromo-hexyl-isoindole-1,3-dione, 1h-isoindole-1,3 2h-dione,2-6-bromohexyl, 1h-isoindole-1,3 2h-dione, 2-6-bromohexyl, 6-n-phthalimido hexyl bromide |
Solubility Information | Insoluble in water. |
Formula Weight | 310.19 |
Percent Purity | 97% |
Chemical Name or Material | N-(6-Bromohexyl)phthalimide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'