missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford N-Acetyl-D-glucosamine
Click to view available options
Quantity:
100 g
1 kg
Unit Size:
100g
1kg
Description
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.
CAS Number: 7512-17-6
Molecular Formula: C8H15NO6
Molecular Weight: 221.21
Specifications
Specifications
| CAS | 7512-17-6 |
| Molecular Formula | C8H15NO6 |
| Quantity | 100 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction