missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford N-Acetyl-D-glucosamine
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.
Brand: Melford A90510-1000.0
This item is not returnable.
View return policy
Description
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.
CAS Number: 7512-17-6
Molecular Formula: C8H15NO6
Molecular Weight: 221.21
Specifications
7512-17-6 | |
C8H15NO6 | |
1 kg |